Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dichloropyridin-2-yl)methanol 97%

Supplier: AMBEED, INC
Warning

A125055-10G A125055-1G
76822-652EA 16.98 USD InStock
76822-652 76822-654 76822-656
(3,5-Dichloropyridin-2-yl)methanol 97%
(3,5-Dichloropyridin-2-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₅Cl₂NO
MDL Number: MFCD08448149
CAS Number: 275383-87-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty