Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-(tert-butyl)-1-methoxybenzene 98%

Supplier: AMBEED, INC
Warning

A125512-1G A125512-250MG A125512-5G
76822-678EA 28 USD InStock
76822-678 76822-680 76822-682
2-Bromo-4-(tert-butyl)-1-methoxybenzene 98%
2-Bromo-4-(tert-butyl)-1-methoxybenzene
Precaución: for Research Use Only


Formula: C₁₁H₁₅BrO
MDL Number: MFCD17214261
CAS Number: 41280-65-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty