Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-1-(methoxymethoxy)-4-methylbenzene 98%

Supplier: AMBEED, INC
Warning

A127396-100MG A127396-1G A127396-250MG
76822-788EA 10 USD InStock
76822-788 76822-790 76822-792
2-Bromo-1-(methoxymethoxy)-4-methylbenzene 98%
2-Bromo-1-(methoxymethoxy)-4-methylbenzene
Precaución: for Research Use Only


Formula: C₉H₁₁BrO₂
MDL Number: MFCD11850106
CAS Number: 104750-60-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty