Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hydroxy-4-propylfuran-2(5H)-one 98%

Supplier: AMBEED, INC
Warning

A128277-100G A128277-10G A128277-1G A128277-25G A128277-5G
76822-826EA 193 USD InStock
76822-826 76822-828 76822-830 76822-832 76822-834
5-Hydroxy-4-propylfuran-2(5H)-one 98%
5-Hydroxy-4-propylfuran-2(5H)-one
Precaución: for Research Use Only


Formula: C₇H₁₀O₃
MW: 142.15 g/mol
MDL Number: MFCD11977490
CAS Number: 78920-10-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty