Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium cinnamate 99%

Supplier: AMBEED, INC
Warning

A130165-100G A130165-10G A130165-1G A130165-25G A130165-500G A130165-5G
76822-968EA 23 USD InStock
76822-968 76822-970 76822-972 76822-974 76822-976 76822-978 76822-980
Potassium cinnamate 99%
Potassium cinnamate
Precaución: for Research Use Only


Formula: C₉H₇KO₂
MDL Number: MFCD00064993
CAS Number: 16089-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty