Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((Trimethylsilyl)ethynyl)phenol 95%

Supplier: AMBEED, INC
Warning

A138092-100MG A138092-250MG
76823-312EA 20 USD InStock
76823-312 76823-314
4-((Trimethylsilyl)ethynyl)phenol 95%
4-((Trimethylsilyl)ethynyl)phenol
Precaución: for Research Use Only


Formula: C₁₁H₁₄OSi
MDL Number: MFCD20257321
CAS Number: 88075-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty