Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-1H-pyrrole 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A140578-100MG A140578-1G A140578-250MG A140578-5G
76823-414EA 106 USD InStock
76823-414 76823-416 76823-418 77683-228
2-Phenyl-1H-pyrrole 98%, Ambeed.Inc
2-Phenyl-1H-pyrrole
Precaución: For research use only.


Formula: C₁₀H₉N
MDL Number: MFCD00963556
CAS Number: 3042-22-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty