Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chlorobenzo[d]thiazole-2-thiol 97%

Supplier: AMBEED, INC
Warning

A140733-10G A140733-1G A140733-250MG
76823-424EA 75 USD InStock
76823-424 76823-426 76823-428
6-Chlorobenzo[d]thiazole-2-thiol 97%
6-Chlorobenzo[d]thiazole-2-thiol
Precaución: for Research Use Only


Formula: C₇H₄ClNS₂
MW: 201.7 g/mol
MDL Number: MFCD00665746
CAS Number: 51618-29-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty