Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(tert-Butoxy)benzeneboronic acid, pinacol ester 97%

Supplier: AMBEED, INC
Warning

A141282-100MG A141282-1G A141282-250MG A141282-5G
76823-446EA 10 USD InStock
76823-446 76823-448 76823-450 76823-452
4-(tert-Butoxy)benzeneboronic acid, pinacol ester 97%
4-(tert-Butoxy)benzeneboronic acid, pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₂₅BO₃
MW: 276.179 g/mol
MDL Number: MFCD08741438
CAS Number: 938063-51-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty