Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6,6-Dimethyl-3-azabicyclo[3.1.0]hexane 97%

Supplier: AMBEED, INC
Warning

A141449-100G A141449-100MG A141449-10G A141449-1G A141449-250MG A141449-25G A141449-5G
76823-462EA 125 USD InStock
76823-462 76823-464 76823-466 76823-468 76823-470 76823-472 76823-474
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane 97%
6,6-Dimethyl-3-azabicyclo[3.1.0]hexane
Precaución: for Research Use Only


Formula: C₇H₁₃N
MW: 111.19 g/mol
MDL Number: MFCD13176114
CAS Number: 943516-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty