Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (R)-oxirane-2-carboxylate 97%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A144615-100MG A144615-250MG A144615-1G
76823-594EA 16 USD InStock
76823-594 76823-596 77683-242
Potassium (R)-oxirane-2-carboxylate 97%, Ambeed.Inc
Potassium (R)-oxirane-2-carboxylate
Precaución: For research use only.


Formula: C₃H₃KO₃
MDL Number: MFCD00274194
CAS Number: 82044-23-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty