Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-2,5-difluorobenzoate 98%

Supplier: AMBEED, INC
Warning

A147552-100G A147552-1G A147552-5G
76823-706EA 439 USD InStock
76823-706 76823-708 76823-710
Methyl 4-bromo-2,5-difluorobenzoate 98%
Methyl 4-bromo-2,5-difluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₅BrF₂O₂
MDL Number: MFCD23713110
CAS Number: 1193162-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty