Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Chloro-3,4-dihydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A148214-100G A148214-10G A148214-25G A148214-500G A148214-5G
76823-732EA 68 USD InStock
76823-732 76823-734 76823-736 76823-738 76823-740
ω-Chloro-3,4-dihydroxyacetophenone 98%
ω-Chloro-3,4-dihydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₇ClO₃
MDL Number: MFCD00002200
CAS Number: 99-40-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty