Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine-3-sulfonic acid 97%

Supplier: AMBEED, INC
Warning

A152485-1G A152485-250MG
76823-866EA 38 USD InStock
76823-866 76823-868
Imidazo[1,2-a]pyridine-3-sulfonic acid 97%
Imidazo[1,2-a]pyridine-3-sulfonic acid
Precaución: for Research Use Only


Formula: C₇H₆N₂O₃S
MW: 198.2 g/mol
MDL Number: MFCD30729948
CAS Number: 112581-51-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty