Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A154744-1G A154744-250MG
76823-942EA 54.11 USD InStock
76823-942 76823-944 76823-946
3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid pinacol ester 98%
3-(Methoxycarbonyl)-5-(trifluoromethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₅H₁₈BF₃O₄
MW: 330.1072 g/mol
MDL Number: MFCD12026075
CAS Number: 1150271-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty