Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-amino-5-chloropyridine-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A161314-100MG A161314-1G A161314-250MG
76824-088EA 14 USD InStock
76824-088 76824-090 76824-092
Methyl 2-amino-5-chloropyridine-4-carboxylate 98%
Methyl 2-amino-5-chloropyridine-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
MW: 186.596 g/mol
MDL Number: MFCD22375099
CAS Number: 1227002-03-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty