Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 5-bromo-2-methoxyisonicotinate 97%

Supplier: AMBEED, INC
Warning

A163937-100MG A163937-250MG
76824-194EA 20 USD InStock
76824-194 76824-196
Methyl 5-bromo-2-methoxyisonicotinate 97%
Methyl 5-bromo-2-methoxyisonicotinate
Precaución: for Research Use Only


Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
MDL Number: MFCD07375112
CAS Number: 886365-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty