Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Tri(2-thienyl)benzene 98%

Supplier: AMBEED, INC
Danger

A164991-100MG A164991-1G A164991-250MG A164991-5G
76824-254EA 23 USD InStock
76824-254 76824-256 76824-258 76824-260
1,3,5-Tri(2-thienyl)benzene 98%
1,3,5-Tri(2-thienyl)benzene
Precaución: for Research Use Only


Formula: C₁₈H₁₂S₃
MW: 324.47 g/mol
MDL Number: MFCD02323391
CAS Number: 15509-95-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty