Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide 95%

Supplier: AMBEED, INC
Warning

A167009-10G A167009-1G A167009-25G
76824-342EA 25 USD InStock
76824-342 76824-344 76824-346
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide 95%
N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide
Precaución: for Research Use Only


Formula: C₇H₆Cl₃NO₅
MW: 290.48 g/mol
MDL Number: MFCD00075216
CAS Number: 66065-85-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty