Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetramisole hydrochloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A167438-100G A167438-10G A167438-1G A167438-25G A167438-500G
76824-360EA 51 USD InStock
76824-360 76824-362 76824-364 76824-366 77683-372
Tetramisole hydrochloride 98%, Ambeed.Inc
Tetramisole hydrochloride
Precaución: For research use only.


Formula: C₁₁H₁₃ClN₂S
MDL Number: MFCD00005536
CAS Number: 5086-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty