Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid 96%

Supplier: AMBEED, INC
Warning

A167617-1G A167617-250MG
76824-372EA 355 USD InStock
76824-372 76824-374
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid 96%
(S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-4,4-difluoro-butyric acid
Precaución: for Research Use Only


Formula: C₁₉H₁₇F₂NO₄
MDL Number: MFCD06796145
CAS Number: 467442-21-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty