Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate 97%

Supplier: AMBEED, INC
Warning

A171191-100G A171191-10G A171191-25G A171191-5G
76824-498EA 46 USD InStock
76824-498 76824-500 76824-502 76824-504
Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride monohydrate 97%
Nalpha-Boc-L-arginine hydrochloride monohydrate
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₄O₄·HCl·H₂O
MDL Number: MFCD00063594
CAS Number: 35897-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty