Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Hydroxy-1-phenylpropyl)-4-methylphenol 95%

Supplier: AMBEED, INC
Warning

A171422-100MG A171422-250MG
76824-506EA 176 USD InStock
76824-506 76824-508
2-(3-Hydroxy-1-phenylpropyl)-4-methylphenol 95%
2-(3-Hydroxy-1-phenylpropyl)-4-methylphenol
Precaución: for Research Use Only


Formula: C₁₆H₁₈O₂
MDL Number: MFCD11100714
CAS Number: 851789-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty