Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-L-alanine 98%

Supplier: AMBEED, INC
Warning

A171485-100G A171485-10G A171485-25G A171485-500G A171485-5G
76824-514EA 42 USD InStock
76824-514 76824-516 76824-518 76824-520 76824-522
N-Acetyl-L-alanine 98%
N-Acetyl-L-alanine
Precaución: for Research Use Only


Formula: C₅H₉NO₃
MW: 131.131 g/mol
MDL Number: MFCD00063132
CAS Number: 97-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty