Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methoxy-2-methylbenzofuran 95%

Supplier: AMBEED, INC
Warning

A172425-100MG A172425-1G A172425-250MG
76824-550EA 25 USD InStock
76824-550 76824-552 76824-554
5-Methoxy-2-methylbenzofuran 95%
5-Methoxy-2-methylbenzofuran
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00619033
CAS Number: 13391-27-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty