Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloro-4-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A172927-100MG A172927-1G A172927-250MG
76824-570EA 40 USD InStock
76824-570 76824-572 76824-574
3-Bromo-5-chloro-4-methylpyridine 95%
3-Bromo-5-chloro-4-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MW: 206.47 g/mol
MDL Number: MFCD18415476
CAS Number: 1260010-08-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty