Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2-methoxypyridine-4-carboxylic acid 96%

Supplier: AMBEED, INC
Warning

A175424-100MG A175424-1G A175424-250MG A175424-5G
76824-688EA 10 USD InStock
76824-688 76824-690 76824-692 76824-694
3-Amino-2-methoxypyridine-4-carboxylic acid 96%
3-Amino-2-methoxypyridine-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₈N₂O₃
MDL Number: MFCD18262266
CAS Number: 870997-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty