Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl-5-aminoisophthalate 98%

Supplier: AMBEED, INC
Warning

A175879-100G A175879-10G A175879-25G A175879-500G A175879-5G
76824-720EA 68 USD InStock
76824-720 76824-722 76824-724 76824-726 76824-728
Dimethyl-5-aminoisophthalate 98%
Dimethyl-5-aminoisophthalate
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
MDL Number: MFCD00008435
CAS Number: 99-27-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty