Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Chloro(phenyl)methylene)bis(fluorobenzene) 95%

Supplier: AMBEED, INC
Danger

A177060-1G A177060-250MG
76824-754EA 256 USD InStock
76824-754 76824-756
4,4'-(Chloro(phenyl)methylene)bis(fluorobenzene) 95%
4,4'-(Chloro(phenyl)methylene)bis(fluorobenzene)
Precaución: for Research Use Only


Formula: C₁₉H₁₃ClF₂
MW: 314.75 g/mol
MDL Number: MFCD17019221
CAS Number: 379-54-4
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty