Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Amino-3-methoxypropanoic acid 97%

Supplier: AMBEED, INC
Warning

A183820-100MG A183820-10G A183820-1G A183820-250MG A183820-25G A183820-5G
76825-026EA 9 USD InStock
76825-026 76825-028 76825-030 76825-032 76825-034 76825-036
(S)-2-Amino-3-methoxypropanoic acid 97%
(S)-2-Amino-3-methoxypropanoic acid
Precaución: for Research Use Only


Formula: C₄H₉NO₃
MDL Number: MFCD00145257
CAS Number: 32620-11-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty