Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-3-(3-bromophenyl)propanoic acid 95%

Supplier: AMBEED, INC
Warning

A187786-100MG A187786-1G A187786-250MG A187786-25G A187786-5G
76825-124EA 5 USD InStock
76825-124 76825-126 76825-128 76825-130 76825-132
(R)-2-Amino-3-(3-bromophenyl)propanoic acid 95%
(R)-2-Amino-3-(3-bromophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀BrNO₂
MDL Number: MFCD06659100
CAS Number: 99295-78-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty