Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane 95%

Supplier: AMBEED, INC
Warning

A194575-100MG A194575-1G A194575-250MG A194575-5G
76825-340EA 12 USD InStock
76825-340 76825-342 76825-344 76825-346
(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane 95%
(3-exo)-3-(3-Isopropyl-5-methyl-4H-1,2,4-triazol-4-yl]-8-azabicyclo[3.2.1]octane
Precaución: for Research Use Only


Formula: C₁₃H₂₂N₄
MDL Number: MFCD12912695
CAS Number: 423165-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty