Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-methyl-1,3-benzoxazole-6-carboxylate 98%

Supplier: AMBEED, INC
Warning

A201411-1G A201411-250MG A201411-5G
76825-580EA 35 USD InStock
76825-580 76825-582 76825-584
Methyl 2-methyl-1,3-benzoxazole-6-carboxylate 98%
Methyl 2-methyl-1,3-benzoxazole-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
MDL Number: MFCD00113064
CAS Number: 136663-23-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty