Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%

Supplier: AMBEED, INC
Warning

A202095-10G A202095-1G A202095-25G A202095-5G
76825-590EA 23 USD InStock
76825-590 76825-592 76825-594 76825-596
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester 97%
2-[2-[(6-Chloro-4-pyrimidinyl)oxy]phenyl]-3-methoxy-2-propenoic acid methyl ester
Precaución: for Research Use Only


Formula: C₁₅H₁₃ClN₂O₄
MDL Number: MFCD13194671
CAS Number: 131860-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty