Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine 98%

Supplier: AMBEED, INC
Warning

A202639-100MG A202639-1G A202639-250MG
76825-604EA 20 USD InStock
76825-604 76825-606 76825-608
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine 98%
N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,1,2,2-tetramethylethylenediamine
Precaución: for Research Use Only


Formula: C₃₆H₅₆N₂O₂
MDL Number: MFCD06797077
CAS Number: 351498-10-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty