Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indazole-6-carboxamide 95%

Supplier: AMBEED, INC
Warning

A204957-1G A204957-250MG A204957-5G
76825-696EA 119 USD InStock
76825-696 76825-698 76825-700
1H-Indazole-6-carboxamide 95%
1H-Indazole-6-carboxamide
Precaución: for Research Use Only


Formula: C₈H₇N₃O
MDL Number: MFCD11643880
CAS Number: 906000-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty