Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Momelotinib 98+%

Supplier: AMBEED, INC
Warning

A207449-1MG A207449-50MG A207449-5MG
76825-772EA 42 USD InStock
76825-772 76825-774 76825-776
Momelotinib 98+%
Momelotinib
Precaución: for Research Use Only


Formula: C₂₃H₂₂N₆O₂
MW: 414.5 g/mol
MDL Number: MFCD16038899
CAS Number: 1056634-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty