Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoronicotinonitrile 95%

Supplier: AMBEED, INC
Warning

A209073-100MG A209073-1G A209073-250MG A209073-5G
76825-812EA 25 USD InStock
76825-812 76825-814 76825-816 76825-818
5-Bromo-2-fluoronicotinonitrile 95%
5-Bromo-2-fluoronicotinonitrile
Precaución: for Research Use Only


Formula: C₆H₂BrFN₂
MDL Number: MFCD18258637
CAS Number: 1256821-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty