Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A209292-100MG A209292-250MG A209292-5G
76825-830EA 42 USD InStock
76825-830 76825-832 77683-594
5-Bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid 95%, Ambeed.Inc
5-Bromo-1H-pyrrolo[2,3-b]pyridine-2-carboxylic acid
Precaución: For research use only.


Formula: C₈H₅BrN₂O₂
MDL Number: MFCD17214372
CAS Number: 1222175-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty