Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Indazol-5-yl)methanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A211334-1G A211334-250MG
76825-912EA 164 USD InStock
76825-912 76825-914
(1H-Indazol-5-yl)methanamine hydrochloride 95%
(1H-Indazol-5-yl)methanamine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClN₃
MDL Number: MFCD23135684
CAS Number: 943845-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty