Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

R-BAIBA 95%

Supplier: AMBEED, INC
Warning

A213846-250MG A213846-1G
76825-964EA 33 USD InStock
76825-964 77568-060
R-BAIBA 95%
R-BAIBA
Precaución: For research use only.


Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD01076229
CAS Number: 2140-95-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty