Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(4-Nitrophenyl)ethanol 95%

Supplier: AMBEED, INC
Warning

A218249-10G A218249-250MG A218249-5G
76826-084EA 107 USD InStock
76826-084 76826-086 76826-088
(R)-1-(4-Nitrophenyl)ethanol 95%
(R)-1-(4-Nitrophenyl)ethanol
Precaución: for Research Use Only


Formula: C₈H₉NO₃
MDL Number: MFCD14705971
CAS Number: 58287-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty