Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxybenzyl bromide 95%

Supplier: AMBEED, INC
Danger

A219877-1G A219877-250MG
76826-140EA 77 USD InStock
76826-140 76826-142
5-Bromo-2-methoxybenzyl bromide 95%
5-Bromo-2-methoxybenzyl bromide
Precaución: for Research Use Only


Formula: C₈H₈Br₂O
MDL Number: MFCD07324845
CAS Number: 184970-28-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty