Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LY-2109761 98%

Supplier: AMBEED, INC
Warning

A225920-10MG A225920-1MG A225920-25MG A225920-2MG A225920-5MG
76826-308EA 42 USD InStock
76826-308 76826-310 76826-312 76826-314 76826-316
LY-2109761 98%
LY-2109761
Precaución: for Research Use Only


Formula: C₂₆H₂₇N₅O₂
MDL Number: MFCD12923354
CAS Number: 700874-71-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty