Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitro-4-(trifluoromethoxy)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A227993-100G A227993-1G A227993-25G A227993-5G
76826-374EA 113 USD InStock
76826-374 76826-376 76826-378 76826-380
3-Nitro-4-(trifluoromethoxy)benzoic acid 98%
3-Nitro-4-(trifluoromethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₄F₃NO₅
MDL Number: MFCD13185757
CAS Number: 784-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty