Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N4,N4'-Diphenyl-N4,N4'-bis(9-phenyl-9H-carbazol-3-yl)-[1,1'-biphenyl]-4,4'-diamine 97%

Supplier: AMBEED, INC
Warning

A231145-10G A231145-1G A231145-5G
76826-450EA 98 USD InStock
76826-450 76826-452 76826-454
N4,N4'-Diphenyl-N4,N4'-bis(9-phenyl-9H-carbazol-3-yl)-[1,1'-biphenyl]-4,4'-diamine 97%
N4,N4'-Diphenyl-N4,N4'-bis(9-phenyl-9H-carbazol-3-yl)-[1,1'-biphenyl]-4,4'-diamine
Precaución: for Research Use Only


Formula: C₆₀H₄₂N₄
MDL Number: MFCD16036285
CAS Number: 887402-92-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty