Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-5-iodo-3-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A231642-1G A231642-250MG A231642-5G
76826-464EA 13 USD InStock
76826-464 76826-466 76826-468
2-Chloro-5-iodo-3-nitropyridine 97%
2-Chloro-5-iodo-3-nitropyridine
Precaución: For research use only.


Formula: C₅H₂ClIN₂O₂
MW: 284.44 g/mol
MDL Number: MFCD04038568
CAS Number: 426463-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty