Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorophenyl)-2,2-difluoroacetic acid 98%

Supplier: AMBEED, INC
Warning

A232454-100MG A232454-1G A232454-250MG A232454-5G
76826-496EA 15 USD InStock
76826-496 76826-498 76826-500 76826-502
2-(4-Chlorophenyl)-2,2-difluoroacetic acid 98%
2-(4-Chlorophenyl)-2,2-difluoroacetic acid
Precaución: for Research Use Only


Formula: C₈H₅ClF₂O₂
MDL Number: MFCD11007727
CAS Number: 475301-73-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty