Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,8-Di(thiophen-2-yl)-1,10-phenanthroline 95%

Supplier: AMBEED, INC
Warning

A248584-100MG A248584-250MG
76826-904EA 100 USD InStock
76826-904 76826-906
3,8-Di(thiophen-2-yl)-1,10-phenanthroline 95%
3,8-Di(thiophen-2-yl)-1,10-phenanthroline
Precaución: for Research Use Only


Formula: C₂₀H₁₂N₂S₂
MDL Number: MFCD22417169
CAS Number: 753491-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty