Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl benzo[d]thiazole-7-carboxylate 95%

Supplier: AMBEED, INC
Warning

A253001-100MG A253001-1G A253001-250MG A253001-5G
76827-012EA 23 USD InStock
76827-012 76827-014 76827-016 76827-018
Methyl benzo[d]thiazole-7-carboxylate 95%
Methyl benzo[d]thiazole-7-carboxylate
Precaución: for Research Use Only


Formula: C₉H₇NO₂S
MDL Number: MFCD13190218
CAS Number: 1038509-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty